Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Demonstration Rod-Mounted Pulley

Supplier: VWR International
DEMO-RDP
470110-528EA 105.6 USD InStock
470110-528
Demonstration Rod-Mounted Pulley
Educational Materials Physics Educational Materials Mechanics Learning Activities
This large aluminum pulley is a great alternative for Atwood’s Machine experiments.

  • Brass Axle and Brass Bush
  • Rod Clamps to a Support
  • Rod Length: 135 mm
  • Rod Width: 10 mm dia.
  • For Atwood’s Machine Experiments

The pulley, 100 mm dia., features smooth-running ball bearings for low friction.

It's attached to a 135 x 10 mm rod that can be clamped to a support.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty