Would you like to log in again?
Acrylic Lens and Prism Set
Supplier: United Scientific Supplies
OPSETP3
470110-634KT 91.85 USD InStock
470110-634
Acrylic Lens and Prism Set
Educational Materials Physics Educational Materials Optics Learning Activities
This set is economical, durable, and versatile.
  • Clear Acrylic Elements
  • Variety of Lens and Prisms
  • Economical
  • Housed in a Convenient Plastic Container
  • Durable
Housed in a convenient plastic container, this set includes seven clear acrylic elements.
Each elements has one frosted surface that reveals the path of a light beam passing through.
Ordering information: This kit includes a double-convex lens, double-concave lens, semicircular lens, trapezoidal lens, small right-angle prism, equilateral prism, and large right-angle prism.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty