Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Double Thistle Osmosis Apparatus
Supplier: G.S.C. INTERNATIONAL, INC.
1501-9
470110-732KT 53.4 USD InStock
470110-732
Double Thistle Osmosis Apparatus
Educational Materials Biology Educational Materials Cell Biology Learning Activities
Osmosis Basic Experiments Have Never Been Clearer.
  • This easy-to-use set includes battery jar
  • 41cm glass double thistle tube with metal support
  • Membrane and O-ring
Water diffuses through the porous membrane at the bottom of the double thistle tube when it is immersed in the water filled battery jar. The diffusion stops when the levels are equal inside and out. Then see the water rise in the tube as a sugar solution is added.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty