Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
G.S.C. Spark Timers
Supplier: G.S.C. INTERNATIONAL, INC.
G.S.C.
TKTMR-1 TKTMR TKTMR-2
470110-812EA 5.05 USD InStock
470110-812 470352-698 470110-810
G.S.C. Spark Timers
Timers Mechanical Timers
The Go Science Crazy spark timer is a recording device used to measure the velocity and acceleration of a falling or moving object.
  • Spark timer device
  • 60 Hz (mains power frequency)
  • Tape length: 45 m
  • Tape width: 17 mm
The Go Science Crazy spark timer sparks at uniform intervals, leaving marks on a paper tape. The distance between the marks on the tape are used to calculate velocity and acceleration. It is provided with a roll of tape, a carbon disc, a weight, and a clamp to hold it in position.
Everything on the spark timer is enclosed for safety.
Delivery information: Timer includes five carbon disks, one 45 m roll of tape, clamp, and weight. Additional rolls of tape and carbon disks are also available separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty