Would you like to log in again?
Center of Mass Apparatus
Supplier: THE SCIENCE SOURCE SE
18650
470110-826EA 76.3 USD InStock
470110-826
Center of Mass Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
Illustrates How Two Bodies Rotate Freely About their Common Center of Mass
  • Includes rod with mounted spheres, wooden handle with pin and instructions
A 54cm rod supports two spheres of different mass at its ends. The rod has two poles, one at its geometric center and one at the model’s center of mass. Mounted at one hole the system spins freely; at the other it does not.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty