Would you like to log in again?
Large Overflow Can
Supplier: United Scientific Supplies
OFLCN1
470110-882EA 24.85 USD InStock
470110-882
Large Overflow Can
Educational Materials Physics Educational Materials Mechanics Learning Activities
For measuring the densities of larger objects.
  • Dimensions are 10cm diameter by 13cm high
  • The 3.5cm spout is 1.7cm below the lip, giving a capacity of about 850ml
A seamless drawn aluminum can with a rolled lip for extra strength.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty