Would you like to log in again?
Atmospheric Pressure Cups
Supplier: VWR International
PH0171
470110-932KT 19.15 USD InStock
470110-932
Atmospheric Pressure Cups
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Demonstrate the Force of Air Pressure Without a Vacuum Pump
In this modern variant of a famous 17th-century experiment, students place the edges of two hemisphere cups together and squeeze them, forcing out the air between them. In a tug of war, students try to pull the cups apart-a surprisingly difficult task. Each of the 7.5cm-diameter rubber hemispheres has a two-finger tug handle and two release posts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty