Would you like to log in again?
Molecular Motion Tube
6003/PRINT
470110-946EA 35.45 USD InStock
470110-946
Molecular Motion Tube
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
A smashing demonstration for the kinetic theory of matter. The 12" glass tube contains numerous colored glass particles and a small amount of mercury. Because the tube is sealed off at reduced pressure, the mercury will boil when placed over a moderate heat source. As the tube is heated, the invisible mercury molecules violently agitate the visible glass particles, creating an effective display of random molecular motion.
Warning: This product contains mercury and may not be available for sale in your state. Contact customer service for details.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty