Would you like to log in again?
Solar Cells
Supplier: SOLAR MADE SE
3-500 4-1.5-200
470111-134CS 415.85 USD InStock
470111-134 470111-136
Solar Cells
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Show How Direct/Indirect Sunlight Provides Power
Safe, simple-to-use encapsulated mini-panels utilize solar cells similar to those that power satellites in space.
Sturdy panels will endure handling without the normal breakage and cell damage associated with solar cells. A unique lens pattern allows light to be collected from all angles.
The 6" red/black lead wires are attached.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty