Would you like to log in again?
Dr. Fuel Cell™ Car Sets
Supplier: HELIOCENTRIS GMBH CP
K0354E+360
470111-138KT 566.65 USD InStock
470111-138
Dr. Fuel Cell™ Car Sets
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Teach Basic Physics and Chemistry With this Exciting Car of the Future That Runs on Distilled Water and Sunlight
No science fiction trick, this car is powered by a miniature version of the same type of fuel cell now being tested in full size cars by eight of the world’s major auto manufacturers! Carefully designed to allow students to perform standard curriculum experiments in Physics and Chemistry as well as to investigate the important emerging technology of fuel cells, the Complete Set comes with everything except water and a lamp, and includes a full set of four course books. The heart of the system is a reversible fuel cell using a Proton Exchange Membrane (a PEM fuel cell). The cell generates hydrogen and oxygen by electrolysis then consumes the gasses to drive the wheels. Shining sunlight or a spotlight onto a solar cell produces electricity for the electrolysis process. The four course books include a comprehensive set of Research Notes and each of the other three books contains twelve prepared lesson sessions oriented to Physical Science, Chemistry, and Physics respectively. Topics are correlated to national standards. The Complete Set contains the Hyco car and fuel cell, a solar cell module, a measuring box, connecting cables. Also available is a Demonstration Set including only the Hyco Car (with instructions) and the solar cell module. You need to supply distilled water and sunlight (or a 90W spotlight).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty