Would you like to log in again?
Replacement Accesssories for Spectra of the Elements Kit
Supplier: THE SCIENCE SOURCE SE
15615
470111-390EA 11.45 USD InStock
470111-390
Replacement Accesssories for Spectra of the Elements Kit
Educational Materials Physics Educational Materials Optics Learning Activities
The replacement chemicals kit contains 5g each of calcium chloride, cupric chloride, cupric nitrate, lithium chloride, potassium chloride, potassium nitrate, sodium bicarbonate, sodium chloride, strontium chloride and strontium nitrate.
An alcohol or bunsen burner is required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty