Would you like to log in again?
Bacteria-3 Types Slide
Supplier: United Scientific Supplies
100-06
470111-738EA 7.15 USD InStock
470111-738
Bacteria-3 Types Slide
Slides Prepared Slides
These slides help you compare and contrast bacteria morphology.
  • Clearly Distinguishable Identifying Features
  • Rigorous Quality Control Standards
  • Slides Show Cocci, Bacillus, and Spirillum Bacterial Forms
They demonstrate cocci, bacillus, and spirillum bacterial forms in three separate smears.
The slides pass rigorous quality control standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty