Would you like to log in again?
Combo-Still™ Fractional Distillation Collector
Supplier: VEGA SCIENTIFIC SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WL5261-35
470111-802EA 115.3 USD InStock
470111-802
Combo-Still™ Fractional Distillation Collector
Water Purification Systems Stills
Organic Synthesis in Microscale Without All the Expensive Glassware
  • Perform organic synthesis, refluxing, and distillation in microscale
  • Set is also reusable and contains enough hardware for one lab setup
  • Activity kits that can be used with the apparatus set are available separately
The Combo-Still™ offers an opportunity to demonstrate or teach organic chemistry in a new and inexpensive way. The inner glass vial contains the reactants and the outer glass vial is filled with silicone oil and is used as a heating bath. Very little heat is needed, and only 2-4 mL of reactant is necessary. The apparatus includes an organic-resistant microplate, experiment stand, thermometer, two glass-bead packed columns (one for reflux, the other for distillation) and a vial of boiling chips. An instruction manual is also included. Fractional Distillation Collector consists of a holder that fits into the Organic Resistant Microplate and six receptacles to collect distillate fractions. Just move the receptacles one-by-one underneath the condenser and collect fractions as necessary.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty