Would you like to log in again?
Lab-Aids® Simulated Urinalysis Kit
Supplier: LAB-AIDS INC (KITS) SE
4
470111-834KT 227 USD InStock
470111-834
Lab-Aids® Simulated Urinalysis Kit
Educational Materials Biology Educational Materials Physiology Learning Activities
How do you test urine?
  • Students perform diagnostic lab tests
  • Simulated urine included
  • Time required: one 50 minute class period
Watch any athletic event on TV and the subject of testing for banned substances will always be discussed in some form of fashion. So how do you teach the basics of urinalysis in the classroom? With this kit! Students analyze simulated urine samples to understand the procedures used in urinalysis and the potential applicability of urinalysis as a diagnostic tool. By adding different reagents to urine samples, students observe how each reacts with substances in the urine, providing useful evidence for identifying different substances. Students test simulated urine samples for pH, density, glucose, phosphates and chlorides. Enough reagents are provided for 30 students.
Ordering information: This kit includes a teacher’s guide with SDS, student worksheets and guides, Chemplates with spatula, density tubes, pH color charts, universal Indicator paper and vial, Benedict’s Solution with drop control bottles, chlorine test solution 1 with drop control bottle, phosphate test solution 1 with drop control bottle, Phosphate Test Solution 2 with drop control bottle, Simulated Urine I with drop control bottle, Simulated Urine II with drop control bottle, Simulated Urine III with drop control bottle, Simulated Urine IV with drop control bottle, and Simulated Urine V with Drop control bottle Materials. A hot plate, Bunsen burner, and test tubes are required but not included.
Delivery information: See datasheet for contents information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty