Would you like to log in again?
Electrophoresis Reagent Package with Methylene Blue
Supplier: Edvotek
604
470111-858EA 130.95 USD InStock
470111-858
Electrophoresis Reagent Package with Methylene Blue
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
UltraSpec-agarose (10 g); tris acetate buffer (50X for 5 L); gel loading solution 10X (1 ml); methylene blue plus staining system (for 3 L) components sufficient for 20 standard gels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty