Would you like to log in again?
Hind III
Supplier: Edvotek
716
470111-876EA 69.7 USD InStock
470111-876
Hind III
Enzymes
Includes instructions for rehydration.Recognition Site: 5'-A^AGCTT-3'(1500 units) 150 ul—for 30 DNA digests
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty