Would you like to log in again?
Easygel™ Tryptone Soy Culture Media, Microbiology Laboratories
Supplier: MICROLOGY INC
TSY40 TSY10
470111-896PK 146.45 USD InStock
470111-896 470111-892
Easygel™ Tryptone Soy Culture Media, Microbiology Laboratories
Media for Microbiology Dehydrated Media for Microbiology
This convenient, one-step media starts as a liquid and gels when poured into the provided petri dishes.
  • Pectin-Gel Method
  • Sterile
  • Pre-Mixed
  • No Autoclaving
  • No Condensation
Easygel is a pectin-gel method
No heating is needed, which reduces preparation time in the lab and makes it easy to take samples on field trips.
As with traditional agar methods, you can add your sample to the liquid medium to create pour plates or let the gel harden to make streak plates.
Ordering information: These products contain living or perishable materials. Easygel comes in a variety of types. The pre-treated petri dishes measure 100 x 15 mm. Shelf life is six months.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty