Would you like to log in again?
Lab-Aids® DNA, RNA Protein Synthesis Kit
Supplier: LAB-AIDS INC (KITS) SE
72
470111-912KT 276 USD InStock
470111-912
Lab-Aids® DNA, RNA Protein Synthesis Kit
Educational Materials Biology Educational Materials Genetics Learning Activities
This kit demystifies DNA's and RNA's connection to protein synthesis.
  • Demystify the DNA/RNA connection
  • Model replication, transcription, and translation
  • Time required: one 50 minutes class period
Students construct a model of DNA, replicate it, and use half of the ladder to build messenger RNA (mRNA). Placing mRNA on a model ribosome simulates transcription in the nucleus and the movement of mRNA. Students simulate translation by matching the mRNA and its codons to transfer RNA (tRNA) and its anti-codons.
Amino acids bond to form a protein molecule, and students experience how information stored in DNA is expressed as a finished protein.
Delivery information: See datasheet for contents information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty