Would you like to log in again?
Mineral Structure - Cleavage and Fracture Experimental Kit
Supplier: LAB-AIDS INC (KITS) SE
402
470112-332KT 259 USD InStock
470112-332
Mineral Structure - Cleavage and Fracture Experimental Kit
Educational Materials Earth Science Educational Materials Minerals and Rocks Learning Activities
Excellent activity to follow study of crystal structure, students identify minerals by physical properties of cleavage and fracture. Included are samples which show cleavage in 1, 2, 3, 4 and 6 directions, and exhibit conchoidal and uneven fracture. Students use a goniometer to study cleavage direction and relate properties of the minerals to their commercial uses. Kits contains five specimens each of: biotite mica, calcite, fluorite, halite, hornblende, microcline feldspar and sphalerite; eight specimens each of: obsidian, serpentine; 15 contact goniometers; and 30 student worksheets. Comes with teacher's instruction booklet providing full background information and materials for 30 students.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty