Would you like to log in again?
Denoyer-Geppert® Deluxe Kidney Model
Supplier: DENOYER-GEPPERT SCI CO SE
0147-00
470112-572EA 567.2 USD InStock
470112-572
Denoyer-Geppert® Deluxe Kidney Model
Models Human Biology Models
The deluxe kidney model is hinged for easy access to internal structures.
  • Model Hinged for Easy Access to Internal Structures
  • Wooded Stand Included
  • Details Shown in Deep Relief
  • Thirty-Four Structures Identified in Provided Key
  • Left Kidney: 3 Times Life Size
The left kidney is three times larger than life size and parasagittally sectioned over three-fourths of its length. The model shows the renal artery and vein, a portion of the ureter, cortex, medulla, pyramids, pelvis, major and minor calyces, and the internal blood supply and drainage in deep relief. Nephrons are detailed in contrasting colors. The distal and proximal convoluted tubules are simplified for clarity. Several loops of Henle with numerous glomeruli are also schematically detailed in relief.
34 structures are identified in the accompanying key.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty