Would you like to log in again?
Animal Mitosis Model Set
Supplier: THE SCIENCE SOURCE SE
B103
470112-580KT 1075 USD InStock
470112-580
Animal Mitosis Model Set
Models Molecular Biology Models
This set of eight clear globes shows the phases of mitosis and chromosome division in a single cell.
  • Height: 5¼"
  • Depth: 3¼"
  • Width of Models Seven and Eight: 6"
  • Different-Colored Plastic Construction
All organelles and cell inclusions including nucleus, nucleolus, double-thread chromosomes, centrioles, centromeres, asters, and spindle fibers are represented in each model. Each one is fully molded in different-colored plastic.
The transparency makes three-dimensional details of the process viewable from any angle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty