Would you like to log in again?
Water Pollution Kit
Supplier: LAB-AIDS INC (KITS) SE
19
470112-628EA 311.35 USD InStock
470112-628
Water Pollution Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
  • Collect local water samples
  • Uses single factor tests
  • Tests are qualitative color changes
Help your students gain an awareness of the significance water pollution plays in the environment. These simple qualitative tests of natural and treated water produce reliable results through vivid color changes. The amount of pollutant in the water can be compared by the depth of color. These ten common pollutants and pH determination can be identified in locally collected water: ammonia, chlorine, chromium, copper, cyanide, iron, nitrate, phosphate, silica and sulfide. Material for a total of 50 tests for each of the listed pollutants.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty