Would you like to log in again?
Dip Net
Supplier: LA MOTTE CHEMICAL PROD CO SE
0168
470112-696EA 111.8 USD InStock
470112-696
Dip Net
Samplers Water Samplers
Easy Water Sampling
  • GLOBE Approved
  • Heavy Duty
This heavy duty dip net is designed for the toughest sampling jobs. It is fitted with a canvas bag and attached to a durable metal frame with clamp rings. The bottom of the bag is nylon of approximately 500 micron. A Canvas skirt extends three inches below bag bottom to protect mesh. The net measures 12" at the rim and is 6-1/2" deep.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty