Would you like to log in again?
PCR Based Alu-Human DNA Typing Kit
Supplier: Edvotek
333-A
470112-840EA 129 USD InStock
470112-840
PCR Based Alu-Human DNA Typing Kit
Educational Materials Biology Educational Materials PCR Learning Activities
Using their own hair or cheek cells as sources of DNA, students will isolate human DNA and compare DNA polymorphisms between individuals.
Students use primers for a 300 base pair Alu insertion in chromosome 16 (PV92) to determine their own genotype. Includes materials for 25 PCR reactions and instructions. A thermal cycler, electrophoresis tank, power supply, variable micropipet and tips, UV transilluminator, balance, and microwave or hot plate are required, but not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty