Would you like to log in again?
Vernier® Anemometer
Supplier: VERNIER SOFTWARE & TECH SE
ANM-BTA
470112-970EA 108.99 USD InStock
470112-970
Vernier® Anemometer
Anemometers
Measure wind speed in dynamic environments.
  • Fits in your palm
  • Standard camera mount
The Vernier Anemometer is an impeller-type anemometer that measures wind speed in the range of 0.5 to 30 m/s (1 to 67 mph). The Anemometer fits in your palm for wind study measurements in the field. A standard camera mount on the back and an accessory rod allows you to position it in wind tunnels or in front of fans for wind turbine experiments.
For educational use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty