Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Basic Wind Experiment Kit (A0018)

Supplier: VERNIER SOFTWARE & TECH SE
KW-BWX
470113-004EA 133.99 USD InStock
470113-004
Basic Wind Experiment Kit (A0018)
Educational Materials Physics Educational Materials Mechanics Learning Activities
Explore wind energy affordably and easily.

  • Create blades for wind turbines
  • Design applications for wind power
  • Grades 4 - 12

This kit, one of our most popular, allows young scientists to test a variety of blade designs, generate electricity, and lift weights. The Basic Wind Experiment Kit has all the materials you need to get started understanding wind power. You provide the wind, basic tools, and your imagination. Great for grades 4–12 and groups, as well as individual science fair projects.

Classroom Pack:
The classroom pack option includes 3 turbines and extra hubs, blade consumables, power output devices (for approximately 24 students).

Specifications:
Height: 24 in
Maximum blade diameter: 36 in (with provided materials)
Maximum number of blades: 12
Expected power output: 0–1 watts
Number of different nacelles: 2

Activities and Concepts:
Activities:
Design turbine blades
Generate electricity
Lift weights
Measure power output
Light LEDs
Compare different designs
Explore torque vs. speed
Charge capacitors
Concepts:
Understanding clean energy
Exploring energy transformations
Exploring the relationship between energy and forces
Developing and using physical models
Understanding differences between energy and power
Collecting data with advanced tools
Analyzing and interpreting data
Using mathematics to explain and understand data
Designing controlled experiments
Optimizing designs.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty