Would you like to log in again?
Investigating the Design and Output of Wet Cell Batteries Kit
Supplier: LAB-AIDS INC (KITS) SE
205S
470113-066EA 243.8 USD InStock
470113-066
Investigating the Design and Output of Wet Cell Batteries Kit
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
See How Electrochemical Cells Operate
  • Students gather, analyze, and interpret data
  • Students use evidence to develop descriptions, predictions, and models
  • Requires one to two 50 minute class periods
Introduce students to the concept of energy conversion. Using the unique LAB–AIDS Wet Cell Chamber, students will design and conduct an experiment to determine which combination of different metals work best to convert potential chemical energy into electric energy. Students will rank the metals tested in order of reactivity as well as investigate how various factors such as the direction of the current flow, the distance between the metals and the amount of metal surface area, affect the electrical output of the wet cell.
Ordering information: Materials Include Table Salt, Copper strips, Iron strips, Magnesium strips, Zinc strips, Electric motors, Wet Cell Chambers SEPUP®, Black wire leads with alligator clips, 3% Hydrogen Peroxide solution, dropper bottles, Graduated cups, Plastic cups, Sandpaper, Red wire leads with alligator clips, Plastic spoons, Transparency 1: Sample Procedure and Data Table for Part B, Transparency 2: Metals Activity Chart.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty