Would you like to log in again?
Biofuels: Investigating Ethanol Production and Combustion Kit
Supplier: LAB-AIDS INC (KITS) SE
39S
470113-076EA 292.65 USD InStock
470113-076
Biofuels: Investigating Ethanol Production and Combustion Kit
Educational Materials Chemistry Educational Materials Consumer Chemistry Learning Activities
Students gain insight on a useful alternative energy source with the Biofuels: Investigating Ethanol Production and Combustion Kit.
  • Kit Provides Three Investigations
  • Investigation One: the Distillation of Fermented Stillage to Produce Fuel
  • Investigation Two Compares Energy Released by the Combustion of Ethanol and Kerosene
  • Investigation Three: Students Discuss Pros and Cons of Fossil Fuels and Biofuels
  • Students Gain Insight on Ethanol Production
This kit gives students a thorough look at ethanol and combustion through three different investigations. Investigation One demonstrates the fermentation of cane sugar and corn, the two food sources most commonly used to produce ethanol, and the distillation of the fermented stillage to produce fuel.
Investigation Two examines the energy released by the combustion of ethanol and kerosene and compares their results. In the final investigation, students compare and measure carbon dioxide and particulate matter, the two by-products of the two fuels and discuss the trade-offs of fossil fuels and biofuels as sources of energy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty