Would you like to log in again?
Dehydrated Media
Supplier: CULTURE MEDIA &SUPPLIES INC SE
LD145B LD184A LD145A CD351A LD100A CD228A CD149A CD146A
470113-142EA 151.45 USD InStock
470113-142 470113-156 470113-144 470113-158 470113-060 470113-150 470113-152 470113-052
Dehydrated Media
Media for Microbiology Dehydrated Media for Microbiology
Dehydrated Media is suitable for almost all educational applications.
  • For Use with Plates, Slants, and Broth Media
  • Instructions Included
  • Sealed Bottle
  • Expiration Date on Bottle
It arrives in a sealed bottle with an expiration date and complete instructions for preparation. Once items have shipped, they have three or more years before expiration.
Agar and components are used for plates or slants. Assuming a 20 mL fill for a 100 x 15 mm petri dish, each 114 g bottle will fill at least 100 dishes; each 454 g bottle will fill at least 400. Broth media are used for tube cultures. Assuming a 12 mL fill for a 20 x 75 mm tube, each 114 g bottle will fill at least 1000 tubes; each 454 g bottle will fill at least 2000.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty