Would you like to log in again?
Student Workstations, Single-Faced Units Phenolic Top, Full Cupboard Configuration
Supplier: DIVERSIFIED WOODCRAFTS INC SE
C2744K C2724K C2734K
470117-658EA 12100 USD InStock
470117-658 470117-662 470117-660
Student Workstations, Single-Faced Units Phenolic Top, Full Cupboard Configuration
Furniture Tables and Workbenches
Single faced tables that provide a fully equipped science center from two to eight students.
  • 3/4" phenolic resin surface
  • Constructed of solid oak and oak hardwood veneers
  • The cabinets have locking cupboard
  • The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
The single faced units contain a solid epoxy resin sink along with cold water faucets for every two students. Every unit contains enough gas cocks for each student along with GFI AC duplex electrical receptacles, rod sockets and book compartments. All units contain locks for the drawers and doors. Add 66" in length for each additional two students.
Sink dimensions: 12L×8W×6"H
Certifications: SEFA compliant and MAS Certified Green.
Ordering information: Choose from models with a full cupboard only or a half cupboard and dovetailed drawers. Can be ordered without sink, fixtures and rod sockets. Your choice of 1" epoxy resin or ¾" phenolic resin tops.
Delivery information: Four student single faced workstation includes two sinks, four cold water/gas fixtures, two GFI protected AC duplex receptacles, four rod sockets and four book compartments in the aprons.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty