Would you like to log in again?
Solar-Powered Car
Supplier: United Scientific Supplies
SLRCR1
470117-792EA 45.55 USD InStock
470117-792
Solar-Powered Car
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
This economical demonstration solar car can be powered by a solar cell or a rechargeable battery (AA size, not included).
  • Economical
  • Rechargable
  • Includes two binding posts
Battery can be recharged from the solar cell, demonstrating an electricity storage system for solar power. The solar cell can be tilted to maximize solar input, features an on/off and select function switch, and includes two binding posts to measure output. All working parts are visible through the clear molded housing. Includes activity guide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty