Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Primary and Secondary Coils

Supplier: United Scientific Supplies
PSC001
470117-794EA 102.55 USD InStock
470117-794
Primary and Secondary Coils
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Primary and Secondary Coils are used to investigate electromagnetic induction.

  • Activity Guide Included
  • Molded Handle for Accurate Positioning
  • Primary Coil Length: 100 mm
  • Self-Inductance: 3.1mH
  • Secondary Coil Length: 102 mm

Primary and secondary coils are wound on robust molded formers fitted with shielded banana jacks. The plated iron core has a molded handle for accurate positioning in the coils. The primary coil has 175 turns of 18 AWG insulated copper wire (diameter: 34 mm, length: 100 mm, resistance: 0.43 Ω, self-inductance: ≈3.1mH).

The secondary coil has three available windings with 500, 1000, and 1500 turns of 26 AWG insulated copper wire (diameter: 52 mm, length: 102 mm, resistances: 11.5 Ω; 23.3 Ω; 35.7 Ω, self-inductances ≈ 490mH; 1.97H; 4.43H). Plated iron core is 19 mm in diameter and 165 mm long (including handle). An activity guide is included.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty