Would you like to log in again?
Basic Simple Machines Kit
Supplier: United Scientific Supplies
FSMKIT02-E
470117-796EA 166.25 USD InStock
470117-796
Basic Simple Machines Kit
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Investigate the basic simple machines with this inclusive kit.
  • Includes all the required materials to start right out of the box
  • Activity manual included
Recommended for grades 3 to 8, this set is ideal for introducing students to the concepts of changing force by levers and pulleys, and the notion of mechanical advantage. The manual describes experiments with the three types of levers, single and double pulley systems, and shows the characteristics of each machine. Please note: This product emits laser light. Max. output power<1mW per beam. Wavelength 650nm. Class IIIa laser product.
Set includes: 1 lever with centimeter scale, 2 Pulleys, 1 Spring scale, 2 Weight hangers, 1 Slotted weight set, 1 Base and support rod, 1 Support rod with clamp, 2 Hook Collars, 1 Pulley cord, & 1 Experiment manual.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty