Would you like to log in again?
Periscope
Supplier: United Scientific Supplies
PSCOPE
470117-800EA 11.9 USD InStock
470117-800
Periscope
Educational Materials Physics Educational Materials Optics Learning Activities
Demonstrate how periscopes in submarines work.
  • Sturdy design
  • Mirrors securely held on each end
Inspired by naval periscopes used on submarines, our handheld periscope is designed to show students the reflective qualities of light.  Our version consists of an outer case, with mirrors at each end set parallel to each other at a 45-degree angle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty