Would you like to log in again?
Surface Plasmon Resonance Apparatus
Supplier: United Scientific Supplies
SPRA01
470117-802EA 3277.05 USD InStock
470117-802
Surface Plasmon Resonance Apparatus
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Utilize the power of a spectrometer to investigate surface plasma resonance.
  • Demonstrate the principles of SPR analysis
  • Use a light beam to excite resonant electron oscillations under specific conditions
  • Includes a collimated laser, polarizer, and photodetector
Surface Plasmon Resonance is an analytical technique used for detecting and tracking a wide variety of surface processes such as DNA and protein interactions, drug mechanisms, immunoassays, anodic stripping, corrosion, gas detection, and many more. In this technique, a light beam interacts with the electron plasma in a metal film at a metal-dielectric interface to excite resonant electron oscillations—Plasmon's—under specific conditions. The component of the photon momentum parallel to the metal surface must match the plasmon momentum. This occurs at a specific incidence angle of the light beam, which is highly sensitive to the state of the surface. In particular, any material attached to the surface produces a detectable change in the resonance angle. The resonance is detected by monitoring the intensity of a totally internally reflected laser beam as a function of incidence angle. At resonance, light energy is absorbed and a significant intensity drop is observed.
The apparatus is used together with our 470110-202 Enhanced Student Spectrometer (not included). The spectrometer’s slit, lenses, and eyepiece are replaced by a collimated laser, a polarizer, and a photodetector. An adjustable support plate for the SPR prism fits onto the spectrometer’s table and an electronic unit controls the laser and measures the light output.
The SPR prism consists of a precise semicircular glass block with a thin layer of gold deposited on the flat surface and a plastic pocket for liquid samples fixed behind it (the Kretschmann configuration.)
At the laser beam’s incidence point, an evanescent wave propagates through the gold layer. Surface plasmons are excited at the interface between the gold layer and the liquid sample, diverting energy from the monitored reflected laser beam.
Includes the following: SPR Electronics Unit, 110V, qty=1; Centering reticle for setup, qty=1; Syringe for inserting sample, qty=2; Polarizing filter unit, qty=1; Connecting cables, qty=2; Optical support for prism, qty=1; Light sensor unit, qty=1; Centering pin puck, qty=1; Laser light source unit, qty=1; SPR prisms, qty=2
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty