Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Student Lab Tables, 1" Phenolic Top

Supplier: DIVERSIFIED WOODCRAFTS INC SE
P7104K30N P7604K30N P7304K30N P7204K30N P7604K36N P7204K36N P7104K36N
470117-830EA 1200 USD InStock
470117-830 470117-842 470117-834 470117-832 470117-838 470117-828 470117-836
Student Lab Tables, 1" Phenolic Top
Furniture Tables and Workbenches
Aprons are constructed of solid red oak with heavy-duty 13-gauge steel corner braces. 2.25" square solid oak legs, with ⁵/₁₆" hanger bolts installed, are attached to apron with nuts and washers supplied. They are smoothly machined with a chemical resistant, earth-friendly UV finish.

  • Available in 2 heights: sitting height 30", standing height 36"
  • 1" phenolic table surface
  • Equipped with non-skid adjustable glides for precise leveling and for prevention of rocking on uneven floors
  • Black rubber leg boots which offer protection against scuffing and chemical spills

Load capacity: 500 lbs.

Minimal assembly required, legs need to be atttached only.

Certifications: SEFA compliant and MAS Certified Green.

Packaging: Shipped KD with tops attached.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty