Would you like to log in again?
Service Island
Supplier: DIVERSIFIED WOODCRAFTS INC SE
3116K
470117-966EA 4600 USD InStock
470117-966
Service Island
Furniture Tables and Workbenches
Use this island to provide water, electricity, and gas to moveable workstations such as Student Tables or the Student Workstations above. This type of furniture arrangement makes moving tables easy for various types of experiments. Worksurface is 1" solid epoxy resin and island includes a 14" x 10" x 6" solid epoxy sink, two cold water faucet/gas cock combinations, and two GFI duplex receptacles. Cabinet at bottom of the island has a lock. Overall Dimensions: 24"L x 26"W x 36"H.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty