Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Android Tablet

470118-116EA 326.39 USD InStock
470118-116
Android Tablet
This Android tablet has all the basic functionality you need to incorporate technology in your hands-on STEM lessons. Access your favorite mobile science apps, integrate probeware technology, add digital science content to your lab or homework assignments, and so much more. Google Play enables easy access to an unlimited selection of apps available for download, including the Ward's Single Probes and Ward's DataHub probeware apps. Designed with education in mind, this tablet comes preloaded with Intel's Education Package which includes the following titles:
Kno textbooks
Intel® Education Lab Camera by Intellisense
MyScript Notes Mobile* Part of Intel® Education
MyScript Stylus Mobile,* Part of Intel® Education
Intel® Education Media Camera by Intellisense
SPARKvue* Part of Intel® Education
ArtRage* Part of Intel® Education
McAfee Mobile Security, Part of Intel® Education
Classroom Management, Part of Intel® Education
Foxit eReader
Intel® Education Resources

Specifications:
OS Android 4.2 Jelly Bean
CPU Intel®Atom Dual Core Clover Trail + 1.2 GHz
Display 10.1”, 1280 x 800 TN Capacitive Multi Touch
Webcam 0.3 M Front Camera, 2M Back Camera with Micro Lens/Light LEDs
Memory 1 x 1Gb LPDDR2 1066MHz
Storage (GB) 8/16/32 eMMC
Connectivity Wi-Fi 802.11 a/b/g/n + BT 4.0
I/O 1 x DC-in Jack
1 x micro-USB 2.0
1 x Combo Audio Jack
1 x Micro HDMI
Sensors G-sensor, eCompass, Light sensor, Temperature probe (working with the GlobiLab app)
Battery 2 cell Li-Polymer 6300 mAh, Battery Life - 8 hours
Applications Standard Android 4.2 applications
Intel Education suite (I'll provide you with the full list)
DataHub data logging analysis app
Weight (g) 735 g
Dimension (mm)
(W) x (D) x (H) 272.9 x 174.4 x 13.2

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty