Would you like to log in again?
Ward's® Chemistry Safer Sodium Demonstration
Supplier: ALDON CORP SE
AD-CC0266-8KIT
470118-182EA 99.09 USD InStock
470118-182
Ward's® Chemistry Safer Sodium Demonstration
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
This demonstration is a safer way to show the reaction between water and alkali metals.
  • A Safer Way to Show the Reaction Between Water and Alkali Metals
  • Demonstration Displays Density, Chemical Reactions, and pH Indicators
  • Cost and Time Efficient
  • For Grades 5-12
  • Time Requirement: 10 Minutes
In this demonstration, students mix two immiscible liquids, mineral oil and water, in a graduated cylinder. A small amount of sodium in the mixture permeates through the mineral oil and react with the two liquids on the bottom.
The reaction, as gas bubbles, form and rise to the mineral oil layer and then drop back down to the water and react yet again. A pH indicator to the water layer displays the production of a base when sodium and water react.
Certifications: Meets NSES for grades five through 12.
Ordering information: This kit includes sodium, phenolphthalein, and mineral oil. Water, graduated cylinders, and forceps are required but not provided.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty