Would you like to log in again?
3B Scientific® Classic Skull And Cervical Spine
Supplier: AMERICAN 3B SCIENTIFIC SE
1020160
470118-894EA 345.65 USD InStock
470118-894
3B Scientific® Classic Skull And Cervical Spine
Models Human Biology Models
Examine The Interaction Between The Skull And Spine
  • High Quality Original Human Skull Cast
  • Can Be Disassembled Into Skull Cap, Base Of Skull And Mandible
  • 3 Year Manufacturers Warranty
  • Includes 3B Smart Anatomy
This human skull replica is flexibly mounted on a cervical spine. Features also shown are the hindbrain, spinal cord, cervical nerves, vertebral arteries, basilar artery and rear cerebral arteries. Skull can be removed from stand. Highly accurate representation of the fissures, foramina, processes and sutures. Mandible of skull is mounted on a spring to easily demonstrate natural movement.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty