Would you like to log in again?
3B Scientific® Half Lower Jaw
Supplier: AMERICAN 3B SCIENTIFIC SE
1000249
470119-068EA 401 USD InStock
470119-068
3B Scientific® Half Lower Jaw
Models Human Biology Models
The model represents half of the left lower jaw of a young person.
  • 2× life size
  • Removable teeth and jaw sections
  • Three years manufacturers warranty
  • Includes 3B smart anatomy
One section of bone is removable from the half lower jaw to expose the tooth roots, spongiosa, vessels and nerves. Canine and first molar are removable from the half lower jaw, and longitudinally sectioned. Half lower jaw on stand.
Accessories information: For information about 3B smart anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty