Would you like to log in again?
3B Scientific® Denture Models
Supplier: AMERICAN 3B SCIENTIFIC SE
1001247 1001248
470119-074EA 344.25 USD InStock
470119-074 470119-072
3B Scientific® Denture Models
Models Human Biology Models
  • Includes 3B Smart Anatomy
Both of these dentition models feature cutaway jaws showing the arrangement of teeth, roots, nerves, blood vessels and bony tissue. The lower jaw on each is moveable for surface and interior examination.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty