Would you like to log in again?
3B Scientific® Desktop Ear Model
Supplier: AMERICAN 3B SCIENTIFIC SE
1000252
470119-104EA 112.35 USD InStock
470119-104
3B Scientific® Desktop Ear Model
Models Human Biology Models
The Desktop Ear Model is ideal for those on a budget.
  • Small Sized Model for Display or Student Study
  • Three-Year Manufacturer's Warranty
  • Includes 3B Smart Anatomy
It represents the outer, middle, and inner ear.
The ear model is on a base, and the ossicles is encased in resin for protection.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty