Would you like to log in again?
3B Scientific® Prostate Model
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1000319
470119-178EA 127.65 USD InStock
470119-178
3B Scientific® Prostate Model
Models Human Biology Models
This prostate model provides a cross section of the male genital organs showing a healthy prostate with bladder, urethra, testicle, symphysis and rectum.
  • 3 year manufacturers warranty
  • Includes 3B Smart Anatomy
The narrowing of the urethra due to the change of the prostate is made clear via the 4 cross sectional views on the bass of the prostate model. The anatomical structures of the prostate model have simple schematic coloring. Prostate model on base.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty