Would you like to log in again?
3B Scientific® Breast Exam Model
Supplier: AMERICAN 3B SCIENTIFIC SE
1000344
470119-202EA 1020.45 USD InStock
470119-202
3B Scientific® Breast Exam Model
Models Human Biology Models
Very realistic natural casting of three different female breasts in C and D cups, to train and practice breast examination.
  • Magnetic Assembly
  • Compare Lactating And Non-Lactating Breasts
  • 3 Year Manufacturers Warranty
Made of new SKINlike™ high-quality silicone. Displays the skin in finest detail. Benign and malignant tumors in different stages of development are featured in two breasts. A healthy breast is supplied for comparison. Supplied with base and talcum powder.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty