Would you like to log in again?
Simulaids® Birthing Simulator
Supplier: NASCO HEALTHCARE INC. SE
110-173 800-225 110-159 110-180
470119-262EA 77.85 USD InStock
470119-262 470119-270 470119-260 470119-266
Simulaids® Birthing Simulator
Models Human Biology Models
Practice Through All Stages Of The Birthing Process
  • Belly overlay contains a fetus in fluid and can be palpated for position
  • Lifelike pelvic cavity
  • Practice multiple procedures
  • Includes a soft bag for storage and transport
An anatomically correct pelvic model with full term newborn and placenta to give your trainees realistic practice in multiple techniques, and to learn the procedure for emergency childbirth. Disposable umbilical cords with clamps, easily replaceable extra vulva, powder to make simulated blood, a modular pregnant belly overlay with permanently installed fetus for training practice of the Leopold’s maneuver to determine the fetus’ lie by palpating its skull and kneecaps, and a clear abdominal overlay to see positioning are all included. The lifelike pelvic cavity has pronounced pelvic landmarks, located spinal column, angled birth canal, ileum, ischium, sacrum, sacro spinious ligaments, and greater sciatic notch. The unit is capable of demonstrating shoulder distocia, fetal attitude, fetal station, fetal lie, cord prolapse, compound presentation, and cesarean section incision locations can be discussed. The fetus provided is of a realistic newborn size with fontanels and cranial sutures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty