Would you like to log in again?
Gaumard® Birthing Simulator With Birthing And Resuscitation Babies
Supplier: AMERICAN 3B SCIENTIFIC SE
1005815 1012417
470119-274EA 6495 USD InStock
470119-274 470119-272
Gaumard® Birthing Simulator With Birthing And Resuscitation Babies
Models Human Biology Models
A Complete Birthing Experience
  • Automatic birthing system
  • Available with birthing and resuscitation babies, or with birthing baby only
Childbirth simulators have been an integral part of Maternal and Child health programs for more than 40 years. The childbirth simulator provides a  comprehensive teaching system which combines the best of our patient care simulators with the advanced childbirth simulator. It is designed to provide a complete birthing experience before, during and after delivery. The birthing simulator comes with four umbilical cords, four dilating cervices, two umbilical clamps, talcum powder, water soluble silicone lubricant, instruction manual and carrying bag.
The birthing simulator features:
Full size articulating female
Intubatable airway with chest rise
IV arm for meds/fluids
Removable stomach cover
Practice Leopold Maneuvers
Multiple fetal heart sounds
Automatic birthing system
Measure head descent and cervical dilation
Multiple placenta locations
Replaceable dilating cervices
One articulating birthing baby with placenta
One resuscitation baby with intubatable airway and umbilical catheter site ( Available on 470119-272 only).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty