Would you like to log in again?
Laerdal® Blood Pressure Training Arm
Supplier: AMERICAN 3B SCIENTIFIC SE
1017977
470119-354EA 2541.59 USD InStock
470119-354
Laerdal® Blood Pressure Training Arm
Models Human Biology Models
Lifelike, Adult Blood Pressure Training Arm
  • Palpable antecubital pulse
  • Blood pressure trainer with LCD guided operation
  • Systolic, diastolic, heart rate and auscultatory gap are programmable
  • Representation of both systolic and diastolic pressures
  • Indication of gauge reading as pressure is increased or decreased
  • Adjustable volume
This blood pressure training arm is complete with a palpable antecubital pulse. Features of this blood pressure arm include:. Blood Pressure Trainer with LCD guided operation. Systolic, diastolic, heart rate and auscultatory gap are programmable. Representation of both systolic and diastolic pressures. Indication of gauge reading as pressure is increased or decreased. Volume adjustable. Includes blood pressure trainer, cuff, 9-volt battery and carry case. This simulator is a great way to teach and learn the technique of taking a patients blood pressure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty