Would you like to log in again?
Life/form® Blood Pressure Training Arm With External Speaker
Supplier: NASCO HEALTHCARE INC. SE
LF01129EX
470119-356EA 2605.9 USD InStock
470119-356
Life/form® Blood Pressure Training Arm With External Speaker
Models Human Biology Models
Easily Train Students On Vital Sounds
  • Instructor controls pressure, pulse, ausculatory gap and volume
  • Includes speaker system
Often when working with a live subject, pressures are difficult to auscultate, making accurate evaluation of student proficiency almost impossible.
This arm features an embedded speaker plus an electronic control unit. It allows the instructor to preset any combination of systolic and diastolic pressures and to control the pulse rate and volume level. An auscultatory gap can also be added and the five Korotkoff phases can be audibly discerned. The pressures are activated at release valve by the student and can be read directly from the sphygmomanometer.
When using the Blood Pressure Simulator, students are never “approximately” correct – with the preset values they are either right or wrong. The external speaker system allows an entire classroom to auscultate blood pressure sounds, which normally can be heard only by one person via stethoscope. The electronic control unit also features a manual calibration button that allows the unit to be used with any type of sphygmomanometer.
The simulator is supplied with the external speaker, the electronic control unit (4 “AA” batteries included), a sphygmomanometer and a hard carrying case.
Ordering information: A stethoscope is not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty