Would you like to log in again?
Sakamoto I.M. Buttock Injection Model
Supplier: AMERICAN 3B SCIENTIFIC SE
1005394
470119-432EA 9625 USD InStock
470119-432
Sakamoto I.M. Buttock Injection Model
Models Human Biology Models
Easy To Learn I.M Positioning For Injection
  • Audio and visual feedback
  • Practice on one side, and visualize on the other
  • Skin does not show needle marks
This unique model of an human buttock has a special two-in-one function: On the right hand side, the anatomy of the bones, ilium crista, greater trochanter, m. gluteus medius, nerves and veins can be studied through the transparent outer structure. The student can transfer what he has learned about the position of nerves, veins etc. to the left hand side on which intra-muscular injections can be practiced. Anatomical landmarks can be palpated through the soft skin to identify the correct positions for injection. The injection of fluid (water) is possible. Correct injections are confirmed by a green light, if the injection is placed in a wrong position or too deep, a red light will appear and a buzzer will sound.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty